missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ DLX5 Polyclonal Antibody
Beskrivelse
Peptide sequence: KEVTEPEVRM VNGKPKKVRK PRTIYSSFQL AALQRRFQKT QYLALPERAE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.
This gene encodes a member of a homeobox transcription factor gene family similar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation.
Tekniske data
Tekniske data
| Antigen | DLX5 |
| Applikationer | Immunohistochemistry, Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | DLX5 |
| Genadgangsnr. | P56178, P70396 |
| Gene Alias | AI385752; distal-less homeo box 5; distal-less homeobox 5; distal-less homeobox protein 5; DLX-3; DLX5; Homeobox protein DLX-3; homeobox protein DLX-5; RDLX; SHFM1D; split hand/foot malformation type 1 with sensorineural hearing loss; Unknown (protein for MGC:137934) |
| Gensymboler | DLX5 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?