Få mere at vide
Novus Biologicals™ DLL4 Antibody Pair [HRP]
Artikelnummer. 18666271 Shop alle Bio Techne produkter
Mængde:
15 tallerkener
5 tallerkener
Artikelnummer. 18666271
Brand: Novus Biologicals™ NBP2795965Plates
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Polyclonal/Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes an antibody specific for Mouse DLL4.
TRUSTED_SUSTAINABILITY
Tekniske data
Analyseområde 9.375 pg/mL
Assay følsomhed 9.38-600 pg/mL
Assay Sandwich ELISA
Konjugeret HRP
Immunoassay-kitformat Sandwich ELISA Kit
Produkttype Antibody Pairs
Specificitet This solid phase sandwich ELISA utilizes an antibody specific for Mouse DLL4.
Målarter Mouse
Til brug med (applikation) Sandwich ELISA
Gene Alias delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Gen-id (Entrez) 54567
Gen symbol DLL4
Værtsarter Rabbit
Sættets indhold
  • Standard: Each vial contains 25 ng of recombinant mouse DLL4.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse DLL4 monoclonal antibody.
  • Detection Antibody: 0.25 mg/mL rabbit anti-DLL4 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Mængde 5 tallerkener
    Regulatorisk status RUO
    Opbevaringskrav Storage is content dependent.
    Mål DLL4
    Teknik 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.