Learn More
R&D Systems™ DLin-KC2-DMA
Product Code. 30537732
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30537732 10 mg 10mg
1 options
Product Code. 30537732 Supplier R&D Systems™ Supplier No. 8895/10
 Restricted Item
Forventet forsendelse: 23-11-2026
A Tocris product, part of R&D Systems. Ionizable cationic lipid
DLin-KC2-DMA is an ionizable cationic lipid (pKa = 6.7), used in the formulation of lipid nanoparticles (LNPs) encapsulating siRNA or plasmid DNA for non-toxic delivery into cells in vitro and in vivo.
For more information on LNPs and available components see our Lipid Nanoparticles page .
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
Product Line Tocris
Quantity 10 mg