missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
DLC1/DLEC1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06065AF750
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DLC1/DLEC1 Polyclonal specifically detects DLC1/DLEC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| DLC1/DLEC1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DLC1, DLEC1, DLEC1 deleted in lung and esophageal cancer 1, F56 | |
| Produced in rabbits immunized with E. coli-derived Human DLC1/DLEC1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9940 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion