Få mere at vide
DL-Tyrosine, TRC
Artikelnummer. 30132440
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132440 50 mg
30135491 100 mg
30011002 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132440 Leverandør Toronto Research Chemicals Leverandørnr. TRCD52537850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 556-03-6
Molekylær formel C9 H11 N O3
Molekylvægt (g/mol) 181.19
Synonym Tyrosine, DL-Tyrosine, Tyrosine, DL- (8CI), (±)-Tyrosine, 2-Amino-3-(4-hydroxyphenyl)propanoic acid, 2-Amino-3-(4-hydroxyphenyl)propionic acid, 2-Azaniumyl-3-(4-hydroxyphenyl)propanoate, D,L-Tyrosine, DL-4-Hydroxyphenylalanine, DL-p-Tyrosine, NSC 205006, α-Amino-4-hydroxybenzenepropanoic acid
IUPAC navn 2-amino-3-(4-hydroxyphenyl)propanoic acid
SMIL NC(Cc1ccc(O)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale DL-Tyrosine
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)
Mængde 50 mg
Formel vægt 181.0739