Få mere at vide
DL-Serine-2,3,3-d3, TRC
Artikelnummer. 30427011
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30427011 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427011 Leverandør Toronto Research Chemicals Leverandørnr. TRCS270982500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Serine-2,3,3-d3
CAS 70094-78-9
Molekylær formel C3 2H3 H4 N O3
Mængde 500 mg
Synonym Serine-2,3,3-d3 (9CI','ACI), DL-Serine-2,3,3-d3 (ZCI), 2-Amino-3-hydroxypropanoic-2,3,3-d3 acid, DL-Serine-2,3,3-D3
InChI formel InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/i1D2,2D
SMIL [2H]C([2H])(O)C([2H])(N)C(=O)O
IUPAC navn 2-amino-2,3,3-trideuterio-3-hydroxypropanoic acid
Molekylvægt (g/mol) 108.11
Formel vægt 108.0614