Få mere at vide
DL-Proline-2,5,5-d3, TRC
Artikelnummer. 30373553
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30373553 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373553 Leverandør Toronto Research Chemicals Leverandørnr. TRCD7560525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Proline-2,5,5-d3
CAS 784086-28-8
Molekylær formel C5 D3 H6 N O2
Mængde 5 mg
Synonym Proline-2,5,5-d3 (9CI, ACI), DL-Proline-2,5,5-d3, DL-Proline-2,5,5-D3, Pyrrolidine-2,5,5-d3-2-carboxylic acid
InChI formel InChI=1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/i3D2,4D
SMIL [2H]C1([2H])CCC([2H])(N1)C(=O)O
IUPAC navn 2,5,5-trideuteriopyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 118.149
Formel vægt 118.082