missing translation for 'onlineSavingsMsg'
Få mere at vide

DL-Pipecolic Acid-d9, TRC

Artikelnummer. 30420536
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420536 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420536 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4797621MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale DL-Pipecolic Acid-d9
CAS 790612-94-1
Renhedsgrad noter HPLC
Molekylær formel C6 D9 H2 N O2
Mængde 1 mg
Synonym 2-Piperidine-2,3,3,4,4,5,5,6,6-d9-carboxylic Acid, 2-Piperidinecarboxylic Acid-d9, Pipecolic Acid-d9, (RS)-2-Piperidinecarboxylic Acid-d9, (±)-2-Piperidinecarboxylic Acid-d9, (±)-Pipecolic Acid-d9, 2-Carboxypiperidine-d9, 2-Pipecolinic Acid-d9, 2-Piperidinylcarboxylic Acid-d9, DL-2-Piperidinecarboxylic Acid-d9, DL-Pipecolic Acid-d9, Dihydrobaikiane-d9, Hexahydro-2-picolinic Acid-d9, Homoproline-d9, NSC 17125-d9, Pipecolinic Acid-d9, Piperidine-6-carboxylic Acid-d9, Piperolinic Acid-d9, α-Pipecolinic Acid-d9, DL-2-Piperidine-d9-carboxylic Acid, DL-Pipecolic Acid D9, 2-Piperidine-2,3,3,4,4,5,5,6,6-d9-carboxylic acid (9CI), [2H9]Pipecolic acid, Homoproline-d9, 2-Piperidine-d9-carboxylic acid
InChI formel InChI=1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/i1D2,2D2,3D2,4D2,5D
SMIL [2H]C1([2H])NC([2H])(C(=O)O)C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 2,3,3,4,4,5,5,6,6-nonadeuteriopiperidine-2-carboxylic acid
Molekylvægt (g/mol) 138.21
Formel vægt 138.1355
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.