Få mere at vide
DL-Panthenol-d6, TRC
Artikelnummer. 30409663
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30409663 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409663 Leverandør Toronto Research Chemicals Leverandørnr. TRCP18120225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Panthenol-d6
CAS 2714485-18-2
Molekylær formel C9 D6 H13 N O4
Mængde 25 mg
Synonym (±)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethyl-butanamide-d6, (±)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide-d6, alpha,gamma-Dihydroxy-N-(3-hydroxypropyl)-beta,beta-dimethylbutyramide-d6, (±)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide-d6, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide-d6, DL-Pantothenol-d6, DL-Pantothenyl Alcohol-d6
InChI formel InChI=1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/i1D3,2D3
SMIL [2H]C([2H])([2H])C(CO)(C(O)C(=O)NCCCO)C([2H])([2H])[2H]
IUPAC navn 4,4,4-trideuterio-2-hydroxy-3-(hydroxymethyl)-N-(3-hydroxypropyl)-3-(trideuteriomethyl)butanamide
Molekylvægt (g/mol) 211.288
Formel vægt 211.169