Få mere at vide
DL-Methionine Methylsulfonium Chloride, TRC
Artikelnummer. 30138699
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30138699 2.5 g 2.5g
17987334 250 mg -
17947924 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138699 Leverandør Toronto Research Chemicals Leverandørnr. TRCD6899582.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3493-12-7
Molekylær formel C6H14ClNO2S
Molekylvægt (g/mol) 199.7
IUPAC navn (3-amino-3-carboxypropyl)-dimethylsulfanium;chloride
SMIL [Cl-].C[S+](C)CCC(N)C(=O)O
Tekniske data
Kemisk navn eller materiale DL-Methionine Methylsulfonium Chloride
Renhedsgrad noter HPLC
InChI formel InChI=1S/C6H13NO2S.ClH/c1-10(2)4-3-5(7)6(8)9;/h5H,3-4,7H2,1-2H3;1H
Mængde 2.5 g
Formel vægt 199.0434
Procent renhed >95%