missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine ethyl ester hydrochloride, 98+%, Thermo Scientific™

Catalog Number 11401140
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6297-53-6
C7H16ClNO2S
213.72
MFCD00237859
KPWCQEUBJAIORR-UHFFFAOYNA-N
dl-methionine ethyl ester hydrochloride, h-met-oet hcl, c7h15no2s.hcl, h-dl-met-oet hcl, h-dl-met-oet.hcl, dl-methionine ethyl ester hcl, ethyl 2-amino-4-methylthiobutanoate, chloride, ethyl 2-azanyl-4-methylsulfanyl-butanoate hydrochloride, 2-amino-4-methylthio butanoic acid ethyl ester hydrochloride, ethyl 2-amino-4-methylsulfanyl butanoate hydrochloride
12641634
Cl.CCOC(=O)C(N)CCSC
This item is not returnable. View return policy
6297-53-6
C7H16ClNO2S
213.72
MFCD00237859
KPWCQEUBJAIORR-UHFFFAOYNA-N
dl-methionine ethyl ester hydrochloride, h-met-oet hcl, c7h15no2s.hcl, h-dl-met-oet hcl, h-dl-met-oet.hcl, dl-methionine ethyl ester hcl, ethyl 2-amino-4-methylthiobutanoate, chloride, ethyl 2-azanyl-4-methylsulfanyl-butanoate hydrochloride, 2-amino-4-methylthio butanoic acid ethyl ester hydrochloride, ethyl 2-amino-4-methylsulfanyl butanoate hydrochloride
12641634
Cl.CCOC(=O)C(N)CCSC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 75°C
IUPAC Name ethyl 2-amino-4-methylsulfanylbutanoate;hydrochloride
Formula Weight 213.73
Percent Purity ≥98%
Quantity 5 g
Sensitivity Hygroscopic
Chemical Name or Material DL-Methionine ethyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'