Få mere at vide
DL-Glyceraldehyde, TRC
Artikelnummer. 30136194
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136194 500 mg
30136536 100 mg
30010895 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136194 Leverandør Toronto Research Chemicals Leverandørnr. TRCG598215500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56-82-6
Molekylær formel C3H6O3
Molekylvægt (g/mol) 90.08
Synonym (±)-2,3-Dihydroxy-propanal, (±)-Glyceraldehyde, 2,3-Dihydroxypropanal, 2,3-Dihydroxypropionaldehyde, DL-Glyceric aldehyde, Glyceraldehyde, Glyceric aldehyde, Glycerinaldehyde, Glycerose, NSC 67934, dl-Glyceraldehyde, alpha,beta-Dihydroxypropionaldehyde
IUPAC navn 2,3-dihydroxypropanal
SMIL OCC(O)C=O
Tekniske data
Kemisk navn eller materiale DL-Glyceraldehyde
Mængde 500 mg
InChI formel InChI=1S/C3H6O3/c4-1-3(6)2-5/h1,3,5-6H,2H2
Formel vægt 90.0317