Få mere at vide
DL-Glutamic Acid-d5, TRC
Artikelnummer. 30240544
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240544 10 mg
30240812 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240544 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59695710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14341-79-8
Molekylær formel C5 D5 H4 N O4
Molekylvægt (g/mol) 152.16
Synonym (±)-Glutamic Acid-d5, DL-Glu-d5, Glutaminic Acid-d5, NSC 206301-d5, NSC 9967-d5, DL-Glutamic-2,3,3,4,4-d5 Acid
IUPAC navn 2-amino-2,3,3,4,4-pentadeuteriopentanedioic acid
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C([2H])(N)C(=O)O
Tekniske data
Kemisk navn eller materiale DL-Glutamic Acid-d5
InChI formel InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/i1D2,2D2,3D
Mængde 10 mg
Formel vægt 152.0845