missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Glutamic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.
Solubility
Soluble in water (miscible), HCl, ether (sparingly), ethanol (sparingly), and petroleum ether (sparingly).
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.
Solubility
Soluble in water (miscible), HCl, ether (sparingly), ethanol (sparingly), and petroleum ether (sparingly).
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications
Beilstein | 1723799 |
Merck Index | 14,4469 |
Solubility Information | Soluble in water (miscible),HCl,ether (sparingly),ethanol (sparingly),and petroleum ether (sparingly). |
Formula Weight | 147.13 |
Percent Purity | 95% |
Quantity | 25 g |
Chemical Name or Material | DL-Glutamic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'