Få mere at vide
DL-Cystine Dihydrochloride, MP Biomedicals™
Artikelnummer. 11473410 Shop alle MP Biomedicals produkter
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
11473410 1 g
11483410 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11473410 Leverandør MP Biomedicals Leverandørnr. 0210145201
Kemiske identifikatorer
CAS 90350-38-2
Molekylær formel C6H14Cl2N2O4S2
Molekylvægt (g/mol) 313.21
MDL nummer MFCD00084652,MFCD00064228
InChI nøgle HHGZUQPEIHGQST-UHFFFAOYNA-N
Synonym cystine dihydrochloride, cytosine-retha-d-riboside hydrochloride, 3,3'-disulfanediylbis 2-aminopropanoic acid dihydrochloride, cystinhydrochlorid, cystine,dihydrochloride, dl-cystine hci, acmc-209bj2, dl-cystine dihydrochloride, dl-cystine dihydrochloride crystalline, 2-amino-3-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride
PubChem CID 12297018
IUPAC navn 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dihydrochloride
SMIL Cl.Cl.NC(CSSCC(N)C(O)=O)C(O)=O
Tekniske data
Kemisk navn eller materiale DL-Cystine Dihydrochloride
Navn note FTIR
Farve Hvid
Mængde 1 g
Formel vægt 313.33
Fysisk form Pulver