Få mere at vide
DL-Cystine-2,2',3,3,3',3'-d6, TRC
Artikelnummer. 30354139
Skift visning
Mængde:
1 mg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354139 2 mg
30363383 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354139 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9978512MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 352431-53-9
Molekylær formel C6 D6 H6 N2 O4 S2
Molekylvægt (g/mol) 246.34
Synonym Cystine-2,2′,3,3,3′,3′-d6 (9CI)
IUPAC navn 2-amino-3-[(2-amino-2-carboxy-1,1,2-trideuterioethyl)disulfanyl]-2,3,3-trideuteriopropanoic acid
SMIL [2H]C([2H])(SSC([2H])([2H])C([2H])(N)C(=O)O)C([2H])(N)C(=O)O
Tekniske data
Kemisk navn eller materiale DL-Cystine-2,2',3,3,3',3'-d6
InChI formel InChI=1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/i1D2,2D2,3D,4D
Mængde 2 mg
Formel vægt 246.0615