Få mere at vide
DL-Cystine-2,2',3,3,3',3'-d6, CDN
Artikelnummer. 30314497
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314497 0.05 g
30313549 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314497 Leverandør CDN Leverandørnr. CDND53010.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Cystine-2,2',3,3,3',3'-d6
CAS 352431-53-9
Molekylær formel C6 D6 H6 N2 O4 S2
Mængde 0.05 g
Synonym Cystine-2,2',3,3,3',3'-d6 (9 CI)
InChI formel InChI=1 S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4 H,1-2,7-8H2,(H,9,10)(H,11,12)/i1D2,2D2,3 D,4 D
SMIL [2 H]C([2 H])(SSC([2 H])([2 H])C([2 H])(N)C(=O)O)C([2 H])(N)C(=O)O
IUPAC navn 2-amino-3-[(2-amino-2-carboxy-1,1,2-trideuterioethyl)disulfanyl]-2,3,3-trideuteriopropanoic acid
Molekylvægt (g/mol) 246.34
Formel vægt 246.0615 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity