Få mere at vide
DL-Cysteine-3,3-d2, TRC
Artikelnummer. 30365550
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30365550 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365550 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99501610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Cysteine-3,3-d2
CAS 124051-54-3
Molekylær formel C3 D2 H5 NO2 S
Mængde 10 mg
Synonym Cysteine-3,3-d2
InChI formel InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/i1D2
SMIL [2H]C([2H])(S)C(N)C(=O)O
IUPAC navn 2-amino-3,3-dideuterio-3-sulfanyl-propanoic acid
Molekylvægt (g/mol) 123.17
Formel vægt 123.0323