Få mere at vide
DL-Alanine-2,3,3,3-d4, CDN
Artikelnummer. 30311796
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311796 1 g -
30321684 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311796 Leverandør CDN Leverandørnr. CDND00921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alanine-d4
CAS 53795-92-9
Molekylær formel C3 D4 H3 N O2
Mængde 1 g
Synonym DL-Alanine-2,3,3,3-d4, DL-Alanine-d4, (R,S)-Alanine-d4, (±)-2-Aminopropionic Acid-d4, (±)-Alanine-d4, DL-Ala-d4, DL-α-Alanine-d4, DL-α-Aminopropionic Acid-d4, NSC 7602-d4, dl-2-Aminopropanoic Acid-d4, dl-Alanine-d4
InChI formel InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/i1D3,2 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(N)C(=O)O
IUPAC navn 2-amino-2,3,3,3-tetradeuteriopropanoic acid
Molekylvægt (g/mol) 93.12
Formel vægt 93.0728 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity