Få mere at vide
DL-4-Hydroxyphenyl-d4-alanine-2,3,3-d3, CDN
Artikelnummer. 30313575
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313575 0.1 g
30311979 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313575 Leverandør CDN Leverandørnr. CDND16900.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Tyrosine-D7
CAS 402835-78-3
Molekylær formel C9 D7 H4 N O3
Mængde 0.1 g
Synonym Tyrosine-α,β,β,2,3,5,6-d7 (9 CI), DL-Tyrosine-α,β,β,2,3,5,6-d7, DL-Tyrosine-D7, 2-Amino-3-(4-hydroxyphenyl-2,3,5,6-d4)propanoic-2,3,3-d3 acid, 4-Hydroxyphenyl-d4-DL-alanine-2,3,3-d3, β-(p-Hydroxyphenyl-d4)-DL-alanine-α,β,β-d3, DL-Tyrosine-d7, DL-4-Hydroxyphenylalanine-d7
InChI formel InChI=1 S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11 H,5,10H2,(H,12,13)/i1D,2 D,3 D,4 D,5D2,8 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)C([2 H])([2 H])C([2 H])(N)C(=O)O
IUPAC navn 2-amino-2,3,3-trideuterio-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 188.2317
Formel vægt 188.1178 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity