Få mere at vide
DL-4-Hydroxyphenyl-2,6-d2-alanine-2-d1, CDN
Artikelnummer. 30320445
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320445 0.1 g 0.1g
30317475 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320445 Leverandør CDN Leverandørnr. CDND16120.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Tyrosine-alpha,2,6-D3
CAS 73036-42-7
Molekylær formel C9 D3 H8 N O3
Mængde 0.1 g
Synonym Tyrosine-α,2,6-d3 (9 CI), DL-Tyrosine-α,2,6-d3, DL-4-Hydroxyphenyl-2,6-d2-alanine-2-d1, DL-Tyrosine-α,2,6-D3, 2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic-2-d acid, DL-Tyrosine-d3, DL-4-Hydroxyphenylalanine-d3
InChI formel InChI=1 S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11 H,5,10H2,(H,12,13)/i1D,2 D,8 D
SMIL [2 H]c1cc(O)cc([2 H])c1CC([2 H])(N)C(=O)O
IUPAC navn 2-amino-2-deuterio-3-(2,6-dideuterio-4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 184.207
Formel vægt 184.0927 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity