Få mere at vide
(DL)-3-O-Methyldopa (3-Methoxy-DL-tyrosine), LoGiCal
Artikelnummer. 30462194
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462194 10 mg
1 options
Artikelnummer. 30462194 Leverandør LoGiCal Leverandørnr. LGCFOR0262.05
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methoxy-DL-tyrosine
CAS 7636-26-2
Analyt- eller komponentnavne 3-Methoxy-DL-tyrosine
Forsendelsestilstand Room Temperature
Molekylær formel C10 H13 N O4
Certificeringer/compliance ISO 9001
Synonym Levodopa Imp. C (EP), Levodopa USP Related Compound B, Levodopa USP RC B, (DL)-3-O-Methyldopa, (2RS)-2-Amino-3-(4-hydroxy-3-methoxyphenyl)propanoic Acid, 3-Methoxy-DL-tyrosine, Levodopa Related Compound B, 3-methoxytyrosine, Levodopa Related Compound B, Levodopa Impurity C
InChI formel InChI=1S/C10H13NO4/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14/h2-3,5,7,12H,4,11H2,1H3,(H,13,14)
Løsningstype Neat
SMIL COc1cc(CC(N)C(=O)O)ccc1O
IUPAC navn 2-amino-3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 211.21
Mængde 10 mg
Formel vægt 211.0845 g/mol