Få mere at vide
DL-2-Aminohexanoic-3,3,4,4,5,5,6,6,6-d9 Acid, CDN
Artikelnummer. 30308880
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30308880 0.1 g 0.1g
30327055 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308880 Leverandør CDN Leverandørnr. CDND79580.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Norleucine-3,3,4,4,5,5,6,6,6-D9
CAS 2243004-88-6
Molekylær formel C6 D9 H4 N O2
Mængde 0.1 g
Synonym 2-Amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid, DL-Norleucine-d9, DL-Norleucine-3,3,4,4,5,5,6,6,6-D9, DL-2-Aminohexanoic-3,3,4,4,5,5,6,6,6-d9 Acid
InChI formel InChI=1 S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/i1D3,2D2,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(N)C(=O)O
IUPAC navn 2-amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid
Molekylvægt (g/mol) 140.2284
Formel vægt 140.1511 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity