Få mere at vide
DL-2-Aminobutyric-2,3,3,4,4,4-d6 Acid, TRC
Artikelnummer. 30314946
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30314946 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314946 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6143212MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminobutanoic-2,3,3,4,4,4-D6 Acid
CAS 350820-17-6
Molekylær formel C4H3D6NO2
Mængde 2 mg
Synonym Butanoic-2,3,3,4,4,4-d6 acid, 2-amino- (9CI), 2-Aminobutanoic-2,3,3,4,4,4-d6 acid, DL-2-Aminobutyric acid-d6, DL-Butyrine-d6, DL-Ethylglycine-d6, DL-Homoalanine-d6, DL-α-Aminobutyric acid-d6, 2-Aminobutanoic acid-d6, 2-Aminobutanoic-2,3,3,4,4,4-D6 acid
InChI formel InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/i1D3,2D2,3D
SMIL O=C(O)C([2H])(N)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn DL-2-Aminobutyric-2,3,3,4,4,4-d6 Acid
Molekylvægt (g/mol) 109.16
Formel vægt 109.1