Få mere at vide
DL-2-Amino-5-phosphonopentanoic Acid, TRC
Artikelnummer. 30240679
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240679 10 mg
30240697 50 mg
30240806 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240679 Leverandør Toronto Research Chemicals Leverandørnr. TRCA62755010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76326-31-3
Molekylær formel C5H12NO5P
Molekylvægt (g/mol) 197.13
Synonym 5-Phosphononorvaline, (±)-2-Amino-5-phosphonopentanoic Acid, (±)-2-Amino-5-phosphonovaleric Acid, 2-APV, 2-Amino-5-phosphonopentanoic Acid, 2-Amino-5-phosphonovaleric Acid, 5-Phosphono-DL-norvaline, AP 5, AP 5 (amino acid), APV, DL-2-Amino-5-phosphonovaleric Acid, DL-AP5
IUPAC navn 2-amino-5-phosphonopentanoic acid
SMIL NC(CCCP(=O)(O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale DL-2-Amino-5-phosphonopentanoic Acid
Mængde 10 mg
InChI formel InChI=1S/C5H12NO5P/c6-4(5(7)8)2-1-3-12(9,10)11/h4H,1-3,6H2,(H,7,8)(H2,9,10,11)
Formel vægt 197.0453