Få mere at vide
Diundecyl Phthalate, TRC
Artikelnummer. 30137635
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137635 25 g
30136394 10 g
17900085 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137635 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49458025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3648-20-2
Molekylær formel C30 H50 O4
Molekylvægt (g/mol) 474.72
Synonym Phthalic Acid, Bis-n-undecyl Ester, 1,2-Benzenedicarboxylic acid, diundecyl ester (9CI), Phthalic acid, diundecyl ester (7CI,8CI), Undecyl alcohol, phthalate (2:1) (8CI), Undecyl alcohol, phthalate (8CI), Bis-diundecylphthalate, Bisoflex DUP, DUP, Di-n-undecyl phthalate, Diplast L 11, Diundecyl phthalate, Jayflex L 11P, Palatinol 11, Sansocizer DUP
IUPAC navn diundecyl benzene-1,2-dicarboxylate
SMIL CCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCC
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis undecyl ester
Mængde 25 g
InChI formel InChI=1S/C30H50O4/c1-3-5-7-9-11-13-15-17-21-25-33-29(31)27-23-19-20-24-28(27)30(32)34-26-22-18-16-14-12-10-8-6-4-2/h19-20,23-24H,3-18,21-22,25-26H2,1-2H3
Formel vægt 474.3709