Få mere at vide
Ditridecyl Phthalate, TRC
Artikelnummer. 30375385
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30375385 10 mg
1 options
Artikelnummer. 30375385 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49409010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(tridecyl) phthalate
CAS 119-06-2
Molekylær formel C34 H58 O4
Mængde 10 mg
Synonym 1,2-Benzenedicarboxylic acid, ditridecyl ester (9CI), Phthalic acid, ditridecyl ester (6CI,7CI,8CI), 1-Tridecanol, phthalate (2:1) (8CI), Bis(tridecyl) phthalate, Di-n-tridecyl phthalate, Ditridecyl phthalate, Hatcol 2922, Hexaplas DTDP, Jayflex DTDP, Polycizer 962BPA, Staflex DTDP, Truflex DTDP
InChI formel InChI=1S/C34H58O4/c1-3-5-7-9-11-13-15-17-19-21-25-29-37-33(35)31-27-23-24-28-32(31)34(36)38-30-26-22-20-18-16-14-12-10-8-6-4-2/h23-24,27-28H,3-22,25-26,29-30H2,1-2H3
SMIL CCCCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCCCC
IUPAC navn ditridecyl benzene-1,2-dicarboxylate
Molekylvægt (g/mol) 530.82
Formel vægt 530.4335