Learn More
Cytiva Dithiothreitol (DTT)
Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. Cytiva Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.
Brand:  Cytiva 17-1318-02
Additional Details : CAS Number : 3483-12-3 Weight : 0.05760kg
 View more versions of this product
Product Code. 11505005
  • 1073.00 DKK / 5g
Estimated Shipment
21-05-2021
Description
Description
  • Use for IEF, SDS-PAGE
  • White
  • Solid
  • Easily soluble in cold water and hot water
  • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
Specifications
Specifications
IEF, SDS-PAGE
0 to 5.1
2000mg
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-dimercaptobutane-2,3-diol