missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithiothreitol, >97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Specifications
Melting Point | 40°C to 43°C |
Boiling Point | 125°C to 130°C/12 mm |
Flash Point | >110°C |
Packaging | Plastic bottle |
Quantity | 5 g |
Beilstein | 1719757 |
Formula Weight | 154.25 |
Percent Purity | ≥97% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Dithiothreitol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'