missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Distilled Water
44.00 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
Description
Gibco™ Distilled Water is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Distilled Water features:
Cell culture quality
Proven performance
Use in cell culture and molecular biology applications
Cell culture quality
Gibco™ Distilled Water is prepared by distillation and filtration (0.1μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.
Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
Use in cell culture and molecular biology applications
Gibco™ Distilled Water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Distilled Water is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Distilled Water product made in our Grand Island facility (15230-170). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Cell culture quality
Gibco™ Distilled Water is prepared by distillation and filtration (0.1μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.
Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
Use in cell culture and molecular biology applications
Gibco™ Distilled Water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Distilled Water is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Distilled Water product made in our Grand Island facility (15230-170). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
Description | Distilled Water, 100mL, Bottle(s), Membrane-Filtered Purification, Sterile, 15 to 30°C Storage Conditions, Room Temperature/Ambient, Fewer Resources Used/Less Waste, ISO 13485/cGMP/FDA Certified |
Sterility | Sterile |
Treatment(s) | Not DEPC-treated |
Certificate of Analysis | ISO 13485, cGMP, FDA |
Quantity | 100 mL |
For Research Use Only. Not for use in diagnostic procedures.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'