Learn More
Lenz Laborglasinstrumente™ Distillation column to Hempel with fixed vacuum jacket 200mm socket/cone 14/23 Lenz
Product Code. 10171951
Change view
Click to view available options
Standard Taper Joint:
14/23
Unit Size:
Each
Product Code. Standard Taper Joint Unit Size
10171951 14/23 Each
1 options
Product Code. 10171951 Supplier Lenz Laborglasinstrumente™ Supplier No. 05119201
  • ONLINE EXCLUSIVE PRICE
    1515.00 DKK
    1126.61 DKK / Pakke med 1
    Spare 388.39 DKK
    25% Slukket
Forventet forsendelse: 25-01-2027