Learn More
Distigmine Bromide, TRC
Product Code. 30387376
Change view
Click to view available options
Quantity:
5 mg
Product Code. Quantity
30387376 5 mg
1 options
Product Code. 30387376 Supplier Toronto Research Chemicals Supplier No. TRCD4935305MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 15876-67-2
Molecular Formula C22 H32 N4 O4 . 2 Br
Quantity 5 mg
Synonym Pyridinium, 3,3'-[1,6-hexanediylbis[(methylimino)carbonyl]oxy]bis[1-methyl-, bromide (1:2), 3-Hydroxy-1-methylpyridinium bromide, hexamethylenebis[methylcarbamate] (6CI,7CI), Pyridinium, 3,3'-[1,6-hexanediylbis[(methylimino)carbonyl]oxy]bis[1-methyl-, dibromide (9CI), Pyridinium, 3-hydroxy-1-methyl-, bromide, hexamethylenebis[methylcarbamate] (2:1) (8CI), BC 51, Distigmine bromide, Distigmine dibromide, Hexamarium, N,N'-Hexamethylenebis(3-N-methylcarbamoxy-1-methylpyridine)dibromide, Ubretid, Ubritil
InChI Formula InChI=1S/C22H32N4O4.2BrH/c1-23-13-9-11-19(17-23)29-21(27)25(3)15-7-5-6-8-16-26(4)22(28)30-20-12-10-14-24(2)18-20;;/h9-14,17-18H,5-8,15-16H2,1-4H3;2*1H/q+2;;/p-2
SMILES [Br-].[Br-].CN(CCCCCCN(C)C(=O)Oc1ccc[n+](C)c1)C(=O)Oc2ccc[n+](C)c2
IUPAC Name (1-methylpyridin-1-ium-3-yl) N-methyl-N-[6-[methyl-(1-methylpyridin-1-ium-3-yl)oxycarbonylamino]hexyl]carbamate;dibromide
Molecular Weight (g/mol) 576.32
Formula Weight 574.079
Chemical Name or Material Distigmine Bromide