missing translation for 'onlineSavingsMsg'
Få mere at vide

Distigmine Bromide, LoGiCal

Artikelnummer. 30457616
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30457616 Leverandør LoGiCal Leverandørnr. LGCFOR1167.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Distigmine Bromide
CAS 15876-67-2
Analyt- eller komponentnavne Distigmine Bromide
Forsendelsestilstand Room Temperature
Molekylær formel C22 H32 N4 O4 . 2 Br
Certificeringer/compliance ISO 9001
Synonym Pyridinium, 3,3'-[1,6-hexanediylbis[(methylimino)carbonyl]oxy]bis[1-methyl-, bromide (1:2), 3-Hydroxy-1-methylpyridinium bromide, hexamethylenebis[methylcarbamate] (6CI,7CI), Pyridinium, 3,3'-[1,6-hexanediylbis[(methylimino)carbonyl]oxy]bis[1-methyl-, dibromide (9CI), Pyridinium, 3-hydroxy-1-methyl-, bromide, hexamethylenebis[methylcarbamate] (2:1) (8CI), BC 51, Distigmine bromide, Distigmine dibromide, Hexamarium, N,N'-Hexamethylenebis(3-N-methylcarbamoxy-1-methylpyridine)dibromide, Ubretid, Ubritil
InChI formel InChI=1S/C22H32N4O4.2BrH/c1-23-13-9-11-19(17-23)29-21(27)25(3)15-7-5-6-8-16-26(4)22(28)30-20-12-10-14-24(2)18-20;;/h9-14,17-18H,5-8,15-16H2,1-4H3;2*1H/q+2;;/p-2
Løsningstype Neat
SMIL [Br-].[Br-].CN(CCCCCCN(C)C(=O)Oc1ccc[n+](C)c1)C(=O)Oc2ccc[n+](C)c2
IUPAC navn (1-methylpyridin-1-ium-3-yl) N-methyl-N-[6-[methyl-(1-methylpyridin-1-ium-3-yl)oxycarbonylamino]hexyl]carbamate;dibromide
Industry Type Clinical Testing
Molekylvægt (g/mol) 576.32
Mængde 10 mg
Formel vægt 574.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.