Learn More
Palintest™ Photometer Tube Test
Predispensed reagent tests require minimal handling and speed up processing. Palintest™; Photometer Tube Test reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Brand: Palintest™ PL 503
Additional Details : Weight : 1.00000kg
Description
- For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
- For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
- Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Specifications
Dissolved Oxygen | |
30 | |
0 to 2.0mg/L |
Photometer, heater block | |
1 Each | |
Photometer tube test reagent |