Learn More
Karl Hecht™ Disposable Volume End-to-End Capillaries
Filling and discharging can be effected with or without a capillary holder.
Brand:  Karl Hecht™ 40551006
 View more versions of this product
Product Code. 10093721
  • 399.00 DKK / Pack of 250
Only null left
Estimated Shipment: 07-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Calibrated to contain from end to end
  • Conformity certified, CE, DIN ISO 7550
  • With capillary holder no. 40553001 (short tube with rubber teat)
  • Accuracy of ≤ ±0.5%
  • Precision of ≤ 1%
  • 250 pieces per pack, 4 packs per carton
Specifications
Specifications
Pipetting
250 per pack; 4 packs per carton
Single-use
Capillary holder no. 40553001
2 ÎĽL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Disposable Volume End-to-End Capillaries > Capacity: 2uL