Få mere at vide
Thermo Scientific™ Disposable IR Cards, 5x10cm, KCl window, 9.5mm aperture
Mængde:
25 pakker
5 pakker
Pakningsstørrelse:
Pakke med 25
Pakke med 5
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Size: 5 x 10cm
- KCl window
- 9.5mm aperture
Tekniske data
Tekniske data
| Oversigt | Disposable IR Cards |
| Mængde | 5 pakker |