Få mere at vide
BRAND™ BLAUBRAND™ IntraEND™ Disposable Micropipets
Artikelnummer. 11864060 Shop alle Brand produkter
Volumen (metrisk):
2 μL
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
Længde (metrisk):
32 mm
61 mm
Pakningsstørrelse:
Pakke med 1000
Artikelnummer. 11864060
Brand: BRAND™ 709109
Comprised of disposable glass without ring mark. BRAND™ Blaubrand™ IntraEND™ Disposable Micropipets are in compliance with DIN ISO 7550 and CE-marked in accordance with IVD-Directive 98/79. Volume is defined by end-to-end filling. Volumes greater than 5μL include DE-M marking.
  • Comprised of disposable glass without ring mark.
  • Complies with DIN ISO 7550.
  • CE Marked in accordance with IVD-Directive 98/79
  • Calibrated to contain (TC, In.)
  • Volume is defined by end-to-end filling
  • Volumes greater than 5μL include DE-M marking.
TRUSTED_SUSTAINABILITY
Tekniske data
Nøjagtighed ≤±0.5%
Kalibrering To Contain
Certificeringer/compliance DIN ISO 7550, IVD-Directive 98/79 CE
Materiale Glass
Mængde 250/Box
Volumen (metrisk) 10 μL
Længde (metrisk) 32 mm
Engangs Single-use