Learn More
HUBER Displacement Insert
For use with refrigeration bath circulators
Brand: HUBER 6293
This item is not returnable.
View return policy
Description
Displacement insertSpecifications
CC-410, CC-410 wl | |
Displacement Insert | |
275 mm |
Product Content Correction
Product Title
HUBER Displacement Insert > Dimensions: 275 x 275 x 189 mm (L x W x H)
Spot an opportunity for improvement?Share a Content Correction