Learn More
HUBER Displacement Insert
For use with refrigeration bath circulators
Brand:  HUBER 6819
 View more versions of this product
Product Code. 10407863
  • 3740.00 DKK / Each
Only null left
Estimated Shipment: 04-08-2025
Description
Displacement insert
Specifications
Specifications
Ministat 230/230 W
Displacement Insert
162 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HUBER Displacement Insert > Dimensions: 162 x 80 x 124 mm (L x W x H)