Learn More
HUBER Displacement Insert
For use with refrigeration bath circulators
Brand: HUBER 6041
This item is not returnable.
View return policy
Description
Displacement insertSpecifications
CC-205B | |
Displacement Insert | |
85 mm |
Product Content Correction
Product Title
HUBER Displacement Insert > Dimensions: 85 x 100 x 133 mm (L x W x H)
Spot an opportunity for improvement?Share a Content Correction