Få mere at vide
Disperse Polyester Dark Blue, 90%, TRC
Artikelnummer. 17920365
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17920365 10 mg
17902835 1 mg
17952385 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17920365 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49346510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56524-76-6
Molekylær formel C16 H14 N2 O4
Molekylvægt (g/mol) 298.29
Synonym 9,10-Anthracenedione, 1,8-dihydroxy-4,5-bis(methylamino)-, 1,8-Dihydroxy-4,5-bis(methylamino)-9,10-anthracenedione, 1,8-Dihydroxy-4,5-bis(methylamino)anthraquinone, Disperse Polyester Dark Blue
IUPAC navn 1,8-dihydroxy-4,5-bis(methylamino)anthracene-9,10-dione
SMIL CNc1ccc(O)c2C(=O)c3c(O)ccc(NC)c3C(=O)c12
Tekniske data
Kemisk navn eller materiale 1,8-Dihydroxy-4,5-bis(methylamino)anthraquinone
Mængde 10 mg
InChI formel InChI=1S/C16H14N2O4/c1-17-7-3-5-9(19)13-11(7)15(21)12-8(18-2)4-6-10(20)14(12)16(13)22/h3-6,17-20H,1-2H3
Formel vægt 298.0954