missing translation for 'onlineSavingsMsg'
Få mere at vide

Disperse Orange 3 (Technical Grade), TRC

Artikelnummer. 30376249
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376249 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30376249 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4934851G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Disperse Orange 3
CAS 730-40-5
Molekylær formel C12 H10 N4 O2
Mængde 1 g
Synonym Benzenamine, 4-[(4-nitrophenyl)azo]- (9CI), C.I. Disperse Orange 3 (6CI,7CI,8CI), 4-[2-(4-Nitrophenyl)diazenyl]benzenamine, 4-(4-Nitrophenylazo)aniline, 4-(4'-Nitrophenylazo)aniline, 4-Amino-4'-nitroazobenzene, 4-Nitro-4'-aminoazobenzene, 4-[(4-Nitrophenyl)azo]benzenamine, 4'-Nitro-4-aminoazobenzene, Acetamine Orange GR, Acetate Fast Orange G, Acetate Fast Orange GR, Acetoquinone Light Orange JRL, Akasperse Orange RBA, Altocyl Orange GR, Amacel Orange GR, Artisil Direct Orange 2R, Artisil Orange 2R, Atlantic Disperse Orange GRA, C.I. 11005, C.I. Solvent Orange 9, Celliton Discharge Orange GRL, Celliton Fast Orange GR, Celliton Orange GR, Celliton Orange GRA, Celutate Orange GRH, Cibacet Orange 2R, Cibacete Orange 2RN, Cilla Fast Orange GR, Diacelliton Fast Orange GL, Disperse Orange 3, Disperse Orange GR, Disperse Orange Zh, Dispersol Fast Orange G, Dispersol Orange AG, Durgacet Orange GR, Eastman Orange GRN, Eastone Orange 2R, Eniacyl Orange GR, Fenacet Orange G, Interchem Acetate Orange GL, Kayalon Fast Orange GR, Microsetile Orange GR, Miketon Fast Orange GR, Nacelan Orange GR, Neosetile Orange GR, Nyloquinone Orange 2R, Oracet Orange 2R, Ostacet Orange P 2R, Perliton Orange 6R, Reliton Orange GR, Serilic Orange GXD, Setacyl Orange GR, Setacyl Orange RE, Silotras Orange TSGR, Supracet Fast Orange G, Synten Orange 2R, Transetile Orange P-GR, Vonteryl Orange GS, p-(p-Nitrophenylazo)aniline, p-Nitro-p'-aminoazobenzene
InChI formel InChI=1S/C12H10N4O2/c13-9-1-3-10(4-2-9)14-15-11-5-7-12(8-6-11)16(17)18/h1-8H,13H2
SMIL Nc1ccc(cc1)N=Nc2ccc(cc2)[N+](=O)[O-]
IUPAC navn 4-[(E)-(4-nitrophenyl)diazenyl]aniline
Molekylvægt (g/mol) 242.23
Formel vægt 242.0804
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.