missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Disperse Orange 29 (Technical Grade), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Disperse Orange 29 |
| CAS | 19800-42-1 |
| Molekylær formel | C19H15N5O4 |
| Mængde | 250 mg |
| Synonym | Phenol, 4-[2-[2-methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]-, Phenol, 4-[[2-methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]- (9CI), Phenol, p-[[2-methoxy-4-[(p-nitrophenyl)azo]phenyl]azo]- (8CI), 4-[2-[2-Methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol, 4-[[2-Methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]phenol, C.I. Disperse Orange 29, Dianix Yellow Brown SE-R, Disperse Orange 29, Foron Yellow Brown SE-RL, Hisperse Orange C-GS, Intrasil Orange L 2R, Intrasil Orange L 2R200, Palanil Orange GL, Resolin Yellow Brown 3GL, Samaron Yellow Brown HRSL, Sumikaron Orange SE-RBL, Synten Orange P-GRL, Phenol, 4-[2-[2-methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]-, Phenol, 4-[[2-methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]- (9CI), Phenol, p-[[2-methoxy-4-[(p-nitrophenyl)azo]phenyl]azo]- (8CI), 4-[2-[2-Methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol, 4-[[2-Methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]phenol, C.I. Disperse Orange 29, Dianix Yellow Brown SE-R, Disperse Orange 29, Foron Yellow Brown SE-RL, Hisperse Orange C-GS, Intrasil Orange L 2R, Intrasil Orange L 2R200, Palanil Orange GL, Resolin Yellow Brown 3GL, Samaron Yellow Brown HRSL, Sumikaron Orange SE-RBL, Synten Orange P-GRL |
| InChI formel | InChI=1S/C19H15N5O4/c1-28-19-12-15(22-20-13-2-7-16(8-3-13)24(26)27)6-11-18(19)23-21-14-4-9-17(25)10-5-14/h2-12,25H,1H3 |
| SMIL | COc1cc(ccc1N=Nc2ccc(O)cc2)N=Nc3ccc(cc3)[N+](=O)[O-] |
| IUPAC navn | 4-[(E)-[2-methoxy-4-[(E)-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol |
| Molekylvægt (g/mol) | 377.35 |
| Formel vægt | 377.1124 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?