Learn More
Miele Professional™ Dosing System
Hose dispenser pump, with level indicator for the automatic dispensing of liquid cleaning agents
Brand:  Miele Professional™ 9968660
Additional Details : Weight : 2.85000kg
 View more versions of this product
Product Code. 15487714
  • 4350.00 DKK / Each
Estimated Shipment
28-09-2020
Add to basket
Description
Description
  • For Miele glass laboratory washer PG 8504
  • Dispenser module for liquid detergent
  • 20cm siphon for canister
  • 'Container empty' indicator
Specifications
Specifications
200 to 230 V
Gray
5 L
200 mm