Få mere at vide
Disopyramide, LoGiCal
Artikelnummer. 30464081
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464081 10 mg
1 options
Artikelnummer. 30464081 Leverandør LoGiCal Leverandørnr. LGCFOR0108.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Disopyramide
CAS 9-5-3737
Analyt- eller komponentnavne Disopyramide
Forsendelsestilstand Room Temperature
Molekylær formel C21 H29 N3 O
Certificeringer/compliance ISO 9001
Synonym Disopyramide
InChI formel InChI=1S/C21H29N3O/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25)
Løsningstype Neat
SMIL CC(C)N(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C(C)C
IUPAC navn 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 339.47
Mængde 10 mg
Formel vægt 339.2311 g/mol