Learn More
Kartell™ Discharge Tube for Dosipet
Kartell™ Discharge Tube for Dosipet is an accessory for Dosipet products, which are used to measure out a variety of solutions and solvents.
129.00 DKK - 166.00 DKK
Specifications
Description Discharge tube
Material FEP
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
13111104
View Documents Promotion Details
Kartell™
8301700
Dosipet 2.5 to 10mL
129.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
13121104
View Documents Promotion Details
Kartell™
8301800
Dosipet 25 to 100mL
166.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
  • For Dosipet products
  • Constructed of FED
  • Good chemical and temperature resistance
Specifications
Specifications
Discharge tube
FEP