Få mere at vide
Dipyridamole EP Impurity A, Mikromol™
Artikelnummer. 30049976
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049976 25 mg
17962016 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049976 Leverandør Mikromol Leverandørnr. MM0462.010025
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dipyridamole EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2'-((4,6,8-Tri(piperidin-1-yl)-pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol
CAS 16982-40-4
Analyt- eller komponentnavne 2,2'-((4,6,8-Tri(piperidin-1-yl)-pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol
Forsendelsestilstand Room Temperature
Molekylær formel C25 H40 N8 O2
Certificeringer/compliance ISO 17025
Synonym 2,2'-[[4,6,8-Tri(piperidin-1-yl)-pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, 2,2'-((4,6,8-Tri(piperidin-1-yl)-pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol, Dipyridamole Related Compound A, Dipyridamole Impurity A
InChI formel InChI=1S/C25H40N8O2/c34-18-16-33(17-19-35)25-27-21-20(23(29-25)31-12-6-2-7-13-31)26-24(32-14-8-3-9-15-32)28-22(21)30-10-4-1-5-11-30/h34-35H,1-19H2
SMIL OCCN(CCO)c1nc(N2CCCCC2)c3nc(nc(N4CCCCC4)c3n1)N5CCCCC5
IUPAC navn 2-[2-hydroxyethyl-[4,6,8-tri(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]amino]ethanol
Molekylvægt (g/mol) 484.64
Mængde 25 mg
Formel vægt 484.3274 g/mol