Få mere at vide
Dipropylene Glycol Diacrylate (~80%), TRC
Artikelnummer. 17972585
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17972585 2.5 g 2.5g
17960545 250 mg -
17940725 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17972585 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4932602.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57472-68-1
Molekylær formel C12H18O5
Molekylvægt (g/mol) 242.27
Synonym Dipropylene Glycol Diacrylate
IUPAC navn 3-(3-prop-2-enoyloxypropoxy)propyl prop-2-enoate
SMIL C=CC(=O)OCCCOCCCOC(=O)C=C
Tekniske data
Kemisk navn eller materiale Dipropylene Glycol Diacrylate
Mængde 2.5 g
InChI formel InChI=1S/C12H18O5/c1-3-11(13)16-9-5-7-15-8-6-10-17-12(14)4-2/h3-4H,1-2,5-10H2
Formel vægt 242.1154