Få mere at vide
Diprenorphine, LoGiCal
Artikelnummer. 30459860
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459860 10 mg
1 options
Artikelnummer. 30459860 Leverandør LoGiCal Leverandørnr. LGCFOR1384.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diprenorphine
CAS 14357-78-9
Analyt- eller komponentnavne Diprenorphine
Forsendelsestilstand Room Temperature
Molekylær formel C26 H35 N O4
Certificeringer/compliance ISO 9001
Synonym 1-N-Cyclopropylmethyl-6,7,8,14-tetrahydro-6,14-endoethano-7-(2'-hydroxy-2'-propyl)nororipavine, Diprenorphine, M 5050, M 5050 Injection, N-(Cyclopropylmethyl)-19-methylnororvinol, RX 5050M, (5alpha,7alpha)-17-(Cyclopropylmethyl)-4,5-epoxy-18,19-dihydro-3-hydroxy-6-methoxy-alpha,alpha-dimethyl-6,14-ethenomorphinan-7-methanol, 21-Cyclopropyl-7alpha-(1-hydroxy-1-methylethyl)-6,14-endo-ethanotetrahydrooripavine
InChI formel InChI=1S/C26H35NO4/c1-23(2,29)18-13-24-8-9-26(18,30-3)22-25(24)10-11-27(14-15-4-5-15)19(24)12-16-6-7-17(28)21(31-22)20(16)25/h6-7,15,18-19,22,28-29H,4-5,8-14H2,1-3H3/t18-,19-,22-,24-,25+,26-/m1/s1
Løsningstype Neat
SMIL CO[C@]12CC[C@@]3(C[C@@H]1C(C)(C)O)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4CC7CC7)c56
IUPAC navn (5α,7α)-17-(Cyclopropylmethyl)-4,5-epoxy-18,19-dihydro-3-hydroxy-6-methoxy-α,α-dimethyl-6,14-ethenomorphinan-7-methanol
Industry Type Clinical Testing
Molekylvægt (g/mol) 425.56
Mængde 10 mg
Formel vægt 425.2566 g/mol