Få mere at vide
Dipotassium Hydrogen Phosphite, TRC
Artikelnummer. 30005138
Skift visning
Mængde:
2.5 g
5 g
25 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30005138 2.5 g 2.5g
30009180 25 g -
30003432 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005138 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4921532.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13492-26-7
Molekylær formel HO3P
Molekylvægt (g/mol) 160.19
SMIL [K+].[K+].[O-]P(=O)[O-]
Tekniske data
Kemisk navn eller materiale Dipotassium Hydrogen Phosphite
Mængde 2.5 g